siRNA Details
| Gene Symbol: Itgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008404.4 | NP_032430.2 | 9 | 1184 | ||
| GenBank mRNA | AK088435.1 | - | 9 | 1173 | ||
| AK136502.1 | - | 9 | 1185 | |||
| AK161236.1 | - | 9 | 1153 | |||
| AK169587.1 | - | 9 | 1219 | |||
| BC145644.1 | - | 8 | 1106 | 40130947 | ||
| M31039.1 | - | 8 | 1072 | |||
| X14951.1 | - | 9 | 1150 | |||

Itgb2
Genomic Map