siRNA Details
| Gene Symbol: Itgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159564.1 | NP_001153036.1 | 9 | 1193 | ||
| NM_021359.3 | NP_067334.1 | 6 | 1087 | |||
| GenBank mRNA | AF115376.1 | - | 9 | 1039 | ||
| AK008867.1 | - | 6 | 1086 | |||
| AK019511.1 | - | 6 | 950 | |||
| AK036439.1 | - | 9 | 1194 | |||
| AK085900.1 | - | 9 | 1234 | |||
| BC049185.1 | - | 6 | 1056 | 6475423 | ||

Itgb6
Genomic Map