siRNA Details
| Gene Symbol: Itih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008406.3 | NP_032432.2 | 11 | 1388 | ||
| GenBank mRNA | AK149500.1 | - | 11 | 1389 | ||
| BC013465.1 | - | 11 | 1372 | |||
| BC028814.1 | - | 11 | 1388 | 5099284 | ||
| X70391.1 | - | 11 | 1368 | |||

Itih1
Genomic Map