siRNA Details
| Gene Symbol: Itih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010582.3 | NP_034712.2 | 10 | 1156 | ||
| GenBank mRNA | AK133782.1 | - | 10 | 1055 | ||
| AK149533.1 | - | 11 | 1201 | |||
| BC034341.1 | - | 10 | 1130 | 4211576 | ||
| X70392.1 | - | 10 | 1142 | |||

Itih2
Genomic Map