siRNA Details
Gene Symbol: Itk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281965.1 | NP_001268894.1 | 13 | 1418 | ||
NM_001281966.1 | NP_001268895.1 | 13 | 1409 | |||
NM_001281968.1 | NP_001268897.1 | 13 | 1397 | |||
NM_010583.3 | NP_034713.2 | 13 | 1400 | |||
GenBank mRNA | AK037554.1 | - | 480 | |||
AK083380.1 | - | 13 | 1372 | |||
AK153613.1 | - | 13 | 1356 | |||
BC128374.1 | - | 13 | 1326 | 40111687 | ||
BC128375.1 | - | 13 | 1323 | 40111688 | ||
D14042.1 | - | 13 | 1340 | |||
L00619.1 | - | 13 | 1405 | |||
L05631.1 | - | 13 | 1366 | |||
L10628.1 | - | 13 | 1368 |
