siRNA Details
| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010584.3 | NP_034714.1 | 2 | 95 | ||
| GenBank mRNA | AB016496.1 | - | 2 | 94 | ||
| AK007447.1 | - | 2 | 95 | |||
| AY157363.1 | - | 2 | 84 | |||
| AY157364.1 | - | 2 | 79 | |||
| AY619693.1 | - | 2 | 78 | |||

Itln1
Genomic Map