siRNA Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013822.5 | NP_038850.1 | 24 | 3237 | ||
| GenBank mRNA | AF171092.1 | - | 24 | 3242 | ||
| AK137092.1 | - | 24 | 3137 | |||
| AK162228.1 | - | 7 | 739 | |||
| BC058675.1 | - | 24 | 3049 | 6834418 | ||

Jag1
Genomic Map