siRNA Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013822.5 | NP_038850.1 | 26 | 3896 | ||
| GenBank mRNA | AF171092.1 | - | 26 | 3901 | ||
| AK078018.1 | - | 3 | 662 | |||
| AK137092.1 | - | 26 | 3796 | |||
| AK162228.1 | - | 9 | 1398 | |||
| BC058675.1 | - | 26 | 3708 | 6834418 | ||

Jag1
Genomic Map