siRNA Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013822.5 | NP_038850.1 | 17 | 2394 | ||
| GenBank mRNA | AF171092.1 | - | 17 | 2399 | ||
| AK137092.1 | - | 17 | 2294 | |||
| BC058675.1 | - | 17 | 2206 | 6834418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jag1| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013822.5 | NP_038850.1 | 17 | 2394 | ||
| GenBank mRNA | AF171092.1 | - | 17 | 2399 | ||
| AK137092.1 | - | 17 | 2294 | |||
| BC058675.1 | - | 17 | 2206 | 6834418 | ||