siRNA Details
Gene Symbol: Jag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010588.2 | NP_034718.2 | 2 | 404 | ||
GenBank mRNA | AF038572.1 | - | 2 | 404 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Jag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010588.2 | NP_034718.2 | 2 | 404 | ||
GenBank mRNA | AF038572.1 | - | 2 | 404 |