siRNA Details
| Gene Symbol: Jak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146145.2 | NP_666257.2 | 7, 8 | 1292 | ||
| GenBank mRNA | AK081552.1 | - | 7, 8 | 1267 | ||
| AK134143.1 | - | 7 | 1235 | |||
| AK141210.1 | - | 7, 8 | 1234 | |||
| AK143650.1 | - | 7, 8 | 1237 | |||
| AK148521.1 | - | 7, 8 | 1257 | |||
| AK152128.1 | - | 7, 8 | 1255 | |||
| BC039752.1 | - | 7, 8 | 1261 | |||
| S63728.1 | - | 7, 8 | 1242 | |||

Jak1
Genomic Map