siRNA Details
| Gene Symbol: Jak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146145.2 | NP_666257.2 | 23 | 3471 | ||
| GenBank mRNA | AK029232.1 | - | 1 | 1731 | ||
| AK133866.1 | - | 23 | 3415 | |||
| AK141210.1 | - | 23 | 3413 | |||
| AK143650.1 | - | 23 | 3418 | |||
| AK152128.1 | - | 23 | 3434 | |||
| AK161759.1 | - | 1 | 1469 | |||
| AK168058.1 | - | 3 | 289 | |||
| BC024100.1 | - | 15 | 1881 | |||
| BC031297.1 | - | 13 | 1592 | |||
| S63728.1 | - | 23 | 3421 | |||

Jak1
Genomic Map