siRNA Details
| Gene Symbol: Jak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146145.2 | NP_666257.2 | 14 | 2260 | ||
| GenBank mRNA | AK133866.1 | - | 14 | 2203 | ||
| AK141210.1 | - | 14 | 2202 | |||
| AK143650.1 | - | 14 | 2207 | |||
| AK148521.1 | - | 14 | 2224 | |||
| AK152128.1 | - | 14 | 2223 | |||
| BC024100.1 | - | 6 | 670 | |||
| BC031297.1 | - | 4 | 381 | |||
| S63728.1 | - | 14 | 2210 | |||

Jak1
Genomic Map