siRNA Details
| Gene Symbol: Jak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190830.1 | NP_001177759.1 | 14 | 2091 | ||
| NM_010589.6 | NP_034719.2 | 14 | 2091 | |||
| GenBank mRNA | AK038268.1 | - | 13 | 1902 | ||
| AK088365.1 | - | 14 | 1940 | |||
| AK142178.1 | - | 14 | 1929 | |||
| AK156646.1 | - | 14 | 1913 | |||
| AK164520.1 | - | 14 | 1931 | |||
| BC105577.1 | - | 13 | 1894 | 3968386 | ||
| BC131646.1 | - | 13 | 1902 | 40110146 | ||
| BC131647.1 | - | 13 | 1902 | 40110148 | ||
| L32955.1 | - | 13 | 1890 | |||
| L33768.1 | - | 11 | 2532 | |||
| L40172.1 | - | 14 | 1952 | |||

Jak3
Genomic Map