siRNA Details
| Gene Symbol: Atcay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178662.3 | NP_848777.1 | 7 | 1031 | ||
| GenBank mRNA | AK028298.1 | - | 7 | 1038 | ||
| AK038483.1 | - | 7 | 1041 | |||
| AK038990.1 | - | 7 | 1032 | |||
| AK039411.1 | - | 7 | 1039 | |||
| AK086723.1 | - | 7 | 1074 | |||
| AK162959.1 | - | 7 | 1039 | |||
| AY349150.1 | - | 7 | 996 | |||
| BC048903.1 | - | 7 | 1015 | 6491141 | ||

Atcay
Genomic Map