siRNA Details
| Gene Symbol: Kcna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008417.5 | NP_032443.3 | 3 | 875 | ||
| GenBank mRNA | AK044342.1 | - | 3 | 877 | ||
| BC138650.1 | - | 2 | 688 | 8861670 | ||
| BC138651.1 | - | 2 | 687 | 8861671 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcna2| Gene Symbol: Kcna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008417.5 | NP_032443.3 | 3 | 875 | ||
| GenBank mRNA | AK044342.1 | - | 3 | 877 | ||
| BC138650.1 | - | 2 | 688 | 8861670 | ||
| BC138651.1 | - | 2 | 687 | 8861671 | ||