siRNA Details
| Gene Symbol: Kcna7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010596.2 | NP_034726.2 | 2 | 1117 | ||
| GenBank mRNA | AF032099.1 | - | 2 | 844 | ||
| AY779767.1 | - | 2 | 851 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcna7| Gene Symbol: Kcna7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010596.2 | NP_034726.2 | 2 | 1117 | ||
| GenBank mRNA | AF032099.1 | - | 2 | 844 | ||
| AY779767.1 | - | 2 | 851 | |||