siRNA Details
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252654.1 | NP_001239583.1 | 11 | 802 | ||
| NM_001252655.1 | NP_001239584.1 | 11 | 831 | |||
| NM_001252656.1 | NP_001239585.1 | 12 | 873 | |||
| NM_010598.3 | NP_034728.2 | 12 | 844 | |||
| GenBank mRNA | AK143166.1 | - | 11 | 779 | ||
| AK157053.1 | - | 11 | 795 | |||
| BC039178.1 | - | 12 | 774 | 4503373 | ||
| L48983.1 | - | 12 | 836 | |||
| U31908.1 | - | 12 | 781 | |||

Kcnab2
Genomic Map