siRNA Details
| Gene Symbol: Kcnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019697.3 | NP_062671.1 | 1 | 1037 | ||
| GenBank mRNA | AB045326.1 | - | 1643 | |||
| AB093280.1 | - | 1 | 1518 | |||
| AF107780.1 | - | 1 | 847 | |||
| AK032268.1 | - | 1828 | ||||
| BC079667.1 | - | 1 | 1037 | 30356567 | ||

Kcnd2
Genomic Map