siRNA Details
| Gene Symbol: Kcnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038607.2 | NP_001033696.1 | 7 | 1248 | ||
| NM_010600.3 | NP_034730.1 | 7 | 1329 | |||
| GenBank mRNA | AK140193.1 | - | 7 | 1235 | ||
| AK147461.1 | - | 7 | 1242 | |||
| BC109012.1 | - | 7 | 1107 | 40048708 | ||
| BC109013.1 | - | 7 | 1188 | 40048709 | ||
| U04294.1 | - | 7 | 1211 | |||

Kcnh1
Genomic Map