siRNA Details
| Gene Symbol: Kcnh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038607.2 | NP_001033696.1 | 6 | 943 | ||
| NM_010600.3 | NP_034730.1 | 6 | 943 | |||
| GenBank mRNA | AK140193.1 | - | 6 | 849 | ||
| AK147461.1 | - | 6 | 937 | |||
| BC109012.1 | - | 6 | 802 | 40048708 | ||
| BC109013.1 | - | 6 | 802 | 40048709 | ||
| U04294.1 | - | 6 | 825 | |||

Kcnh1
Genomic Map