siRNA Details
| Gene Symbol: Kcnj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008425.4 | NP_032451.1 | 2 | 570 | ||
| GenBank mRNA | AF021136.1 | - | 1 | 164 | ||
| AK044915.1 | - | 2 | 571 | |||
| BB642909.1 | - | 2 | 571 | |||
| BC137841.1 | - | 1 | 340 | 8860860 | ||
| BC137842.1 | - | 1 | 340 | 8860861 | ||
| X73052.1 | - | 2 | 501 | |||

Kcnj2
Genomic Map