siRNA Details
| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046778.1 | - | 3 | 1541 | ||
| BC068167.1 | - | 4 | 2419 | 30543982 | ||
| BC103671.1 | - | 3 | 1496 | |||
| BC103672.1 | - | 2 | 1278 | |||
| BC103673.1 | - | 3 | 1495 | 40046971 | ||
| BC103674.1 | - | 3 | 1495 | 40046969 | ||
| D45022.1 | - | 3 | 1503 | |||

Kcnj3
Genomic Map