siRNA Details
| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046778.1 | - | 2, 3 | 1001 | ||
| BC068167.1 | - | 3, 4 | 1880 | 30543982 | ||
| BC103671.1 | - | 2, 3 | 956 | |||
| BC103673.1 | - | 2, 3 | 956 | 40046971 | ||
| BC103674.1 | - | 2, 3 | 956 | 40046969 | ||
| D45022.1 | - | 2, 3 | 964 | |||

Kcnj3
Genomic Map