siRNA Details
| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046778.1 | - | 1 | 325 | ||
| BC068167.1 | - | 2 | 1204 | 30543982 | ||
| BC103671.1 | - | 1 | 280 | |||
| BC103672.1 | - | 1 | 279 | |||
| BC103673.1 | - | 1 | 280 | 40046971 | ||
| BC103674.1 | - | 1 | 280 | 40046969 | ||
| D45022.1 | - | 1 | 288 | |||

Kcnj3
Genomic Map