siRNA Details
| Gene Symbol: Kcnj9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008429.2 | NP_032455.2 | 4 | 1142 | ||
| GenBank mRNA | AF130860.1 | - | 4 | 1105 | ||
| AK033970.1 | - | 4 | 1142 | |||
| AK046084.1 | - | 4 | 1181 | |||
| BC065161.1 | - | 3 | 943 | 5718431 | ||
| U11860.1 | - | 4 | 1191 | |||

Kcnj9
Genomic Map