siRNA Details
| Gene Symbol: Khk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008439.3 | NP_032465.2 | 5 | 453 | ||
| GenBank mRNA | BC013464.1 | - | 5 | 453 | 4235560 | |
| Y09335.1 | - | 5 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khk| Gene Symbol: Khk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008439.3 | NP_032465.2 | 5 | 453 | ||
| GenBank mRNA | BC013464.1 | - | 5 | 453 | 4235560 | |
| Y09335.1 | - | 5 | 432 | |||