siRNA Details
| Gene Symbol: Kif11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010615.1 | NP_034745.1 | 13 | 1721 | ||
| GenBank mRNA | AJ223293.1 | - | 12 | 1416 | ||
| AK050778.1 | - | 2 | 1508 | |||
| AK083324.1 | - | 13 | 1698 | |||
| AK147754.1 | - | 13 | 1745 | |||
| BC060670.1 | - | 13 | 1721 | 6406754 | ||

Kif11
Genomic Map