siRNA Details
| Gene Symbol: Kif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110315.2 | NP_001103785.1 | 11 | 1099 | ||
| NM_001294149.1 | NP_001281078.1 | 11 | 1099 | |||
| NM_001294150.1 | NP_001281079.1 | 10 | 987 | |||
| NM_008440.4 | NP_032466.2 | 11 | 1099 | |||
| GenBank mRNA | AK147640.1 | - | 11 | 1057 | ||
| AY426748.1 | - | 10 | 928 | |||
| BC054538.1 | - | 11 | 1072 | |||
| BC062891.1 | - | 11 | 1078 | 5702372 | ||
| D29951.1 | - | 11 | 1048 | |||

Kif1a
Genomic Map