siRNA Details
Gene Symbol: Kif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110315.2 | NP_001103785.1 | 31 | 3350 | ||
NM_001294149.1 | NP_001281078.1 | 31 | 3350 | |||
NM_001294150.1 | NP_001281079.1 | 31 | 3265 | |||
NM_008440.4 | NP_032466.2 | 31 | 3350 | |||
GenBank mRNA | AK147640.1 | - | 31 | 3308 | ||
AK220487.1 | - | 16 | 1526 | |||
AY426748.1 | - | 31 | 3206 | |||
BC054538.1 | - | 31 | 3322 | |||
BC062891.1 | - | 31 | 3329 | 5702372 | ||
D29951.1 | - | 31 | 3299 |
