siRNA Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153103.2 | NP_694743.2 | 12 | 1350 | ||
| GenBank mRNA | AB074017.1 | - | 10 | 980 | ||
| AK029250.1 | - | 12 | 1277 | |||
| AK046574.1 | - | 7 | 591 | |||
| AK173007.1 | - | 11 | 1057 | |||
| BC029286.1 | - | 12 | 1337 | |||

Kif1c
Genomic Map