siRNA Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153103.2 | NP_694743.2 | 8 | 1030 | ||
| GenBank mRNA | AB001456.1 | - | 4 | 381 | ||
| AB074017.1 | - | 6 | 660 | |||
| AK029250.1 | - | 8 | 957 | |||
| AK046574.1 | - | 3 | 271 | |||
| AK173007.1 | - | 7 | 737 | |||
| BC029286.1 | - | 8 | 1017 | |||

Kif1c
Genomic Map