siRNA Details
| Gene Symbol: Kif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145779.1 | NP_001139251.1 | 19 | 2003 | ||
| NM_008442.2 | NP_032468.2 | 21 | 2500 | |||
| GenBank mRNA | AK037037.1 | - | 19 | 1916 | ||
| BB622253.1 | - | 3 | 273 | |||
| BC006803.1 | - | 19 | 1998 | 3152883 | ||
| D12644.1 | - | 21 | 2501 | |||
| Y15894.1 | - | 18 | 1853 | |||

Kif2a
Genomic Map