siRNA Details
| Gene Symbol: Kif21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039472.1 | NP_001034561.1 | 29, 30 | 4310 | ||
| GenBank mRNA | AF202893.1 | - | 29, 30 | 4288 | ||
| CA328354.1 | - | 3, 4 | 302 | |||
| CK781103.1 | - | 5, 6 | 399 | |||

Kif21b
Genomic Map