siRNA Details
| Gene Symbol: Kif21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039472.1 | NP_001034561.1 | 19 | 3087 | ||
| GenBank mRNA | AF202893.1 | - | 19 | 3065 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif21b| Gene Symbol: Kif21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039472.1 | NP_001034561.1 | 19 | 3087 | ||
| GenBank mRNA | AF202893.1 | - | 19 | 3065 | ||