siRNA Details
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008446.2 | NP_032472.2 | 7 | 1289 | ||
GenBank mRNA | AK084837.1 | - | 7 | 1266 | ||
AK135179.1 | - | 7 | 1237 | |||
AK135430.1 | - | 7 | 783 | |||
AK171938.1 | - | 7 | 1202 | |||
BC050946.1 | - | 7 | 1227 | 6822283 | ||
BC054537.1 | - | 7 | 1211 | |||
D12646.1 | - | 7 | 1616 |
