siRNA Details
| Gene Symbol: Uhmk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010633.3 | NP_034763.3 | 1, 2 | 416 | ||
| GenBank mRNA | AK013347.1 | - | 1, 2 | 421 | ||
| BC058732.1 | - | 1, 2 | 416 | 6414877 | ||
| Y10725.2 | - | 1, 2 | 273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uhmk1| Gene Symbol: Uhmk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010633.3 | NP_034763.3 | 1, 2 | 416 | ||
| GenBank mRNA | AK013347.1 | - | 1, 2 | 421 | ||
| BC058732.1 | - | 1, 2 | 416 | 6414877 | ||
| Y10725.2 | - | 1, 2 | 273 | |||