siRNA Details
| Gene Symbol: Klf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010636.3 | NP_034766.2 | 6 | 939 | ||
| GenBank mRNA | AB221575.1 | - | 6 | 907 | ||
| AK042450.1 | - | 1 | 191 | |||
| BC067408.1 | - | 6 | 939 | 30091275 | ||
| Y14295.1 | - | 6 | 923 | |||

Klf12
Genomic Map