siRNA Details
| Gene Symbol: Kpna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008467.4 | NP_032493.1 | 3 | 477 | ||
| GenBank mRNA | AF020771.1 | - | 3 | 185 | ||
| AK147920.1 | - | 3 | 486 | |||
| BC026821.1 | - | 3 | 272 | 3672708 | ||
| BC052162.1 | - | 3 | 478 | 6510795 | ||
| CT010281.1 | - | 3 | 185 | |||

Kpna4
Genomic Map