siRNA Details
| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010657.3 | NP_034787.2 | 5, 6 | 6028 | ||
| GenBank mRNA | AK173200.1 | - | 2, 3 | 1274 | ||
| AY454345.1 | - | 6, 7 | 5975 | |||
| L07911.1 | - | 2, 3 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hivep3| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010657.3 | NP_034787.2 | 5, 6 | 6028 | ||
| GenBank mRNA | AK173200.1 | - | 2, 3 | 1274 | ||
| AY454345.1 | - | 6, 7 | 5975 | |||
| L07911.1 | - | 2, 3 | 629 | |||