siRNA Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 9 | 1251 | ||
| GenBank mRNA | AK036566.1 | - | 9 | 1307 | ||
| AK137532.1 | - | 8 | 988 | |||
| AK164833.1 | - | 9 | 1373 | |||
| U43585.1 | - | 9 | 1258 | |||
| X81634.1 | - | 9 | 1033 | |||

Ksr1
Genomic Map