siRNA Details
| Gene Symbol: Ktn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293635.1 | NP_001280564.1 | 9 | 1549 | ||
| NM_001293636.1 | NP_001280565.1 | 10 | 1669 | |||
| NM_008477.2 | NP_032503.2 | 8 | 1420 | |||
| GenBank mRNA | AK040200.1 | - | 9 | 1547 | ||
| BC046994.1 | - | 9 | 1549 | |||
| BC066871.1 | - | 9 | 1504 | |||
| BC075697.1 | - | 10 | 1588 | |||
| L43326.1 | - | 8 | 1408 | |||

Ktn1
Genomic Map