siRNA Details
Gene Symbol: Lama2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008481.2 | NP_032507.2 | 37, 38 | 5586 | ||
GenBank mRNA | AK220480.1 | - | 19, 20 | 2806 | ||
CD350015.1 | - | 1, 2 | 87 | |||
U12147.1 | - | 37, 38 | 5494 |
