siRNA Details
| Gene Symbol: Lama5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081171.2 | NP_001074640.1 | 69 | 9677 | ||
| GenBank mRNA | AK129157.1 | - | 30 | 4451 | ||
| AK158204.1 | - | 2 | 304 | |||
| BC020313.2 | - | 6 | 849 | |||
| BC062207.1 | - | 7 | 1110 | |||
| U37501.1 | - | 69 | 9371 | |||

Lama5
Genomic Map