siRNA Details
| Gene Symbol: Lamb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008483.3 | NP_032509.2 | 5 | 538 | ||
| GenBank mRNA | AK140351.1 | - | 5 | 517 | ||
| AK158444.1 | - | 5 | 510 | |||
| AK212573.1 | - | 2 | 58 | |||
| BC026051.1 | - | 5 | 465 | 4036357 | ||

Lamb2
Genomic Map