siRNA Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008485.3 | NP_032511.3 | 24 | 3864 | ||
| GenBank mRNA | DV658346.1 | - | 3 | 491 | ||
| U43327.2 | - | 23 | 3591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamc2| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008485.3 | NP_032511.3 | 24 | 3864 | ||
| GenBank mRNA | DV658346.1 | - | 3 | 491 | ||
| U43327.2 | - | 23 | 3591 | |||