siRNA Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008485.3 | NP_032511.3 | 15 | 2479 | ||
| GenBank mRNA | U43327.2 | - | 14 | 2209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamc2| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008485.3 | NP_032511.3 | 15 | 2479 | ||
| GenBank mRNA | U43327.2 | - | 14 | 2209 | ||