siRNA Details
| Gene Symbol: Anpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008486.2 | NP_032512.2 | 1 | 327 | ||
| GenBank mRNA | AK143746.1 | - | 1 | 270 | ||
| BC005431.1 | - | 1 | 293 | 3593307 | ||
| U77083.1 | - | 1 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anpep| Gene Symbol: Anpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008486.2 | NP_032512.2 | 1 | 327 | ||
| GenBank mRNA | AK143746.1 | - | 1 | 270 | ||
| BC005431.1 | - | 1 | 293 | 3593307 | ||
| U77083.1 | - | 1 | 259 | |||