siRNA Details
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130150.1 | NP_001123622.1 | 23 | 2313 | ||
NM_001130151.1 | NP_001123623.1 | 22 | 2304 | |||
NM_001130152.1 | NP_001123624.1 | 21 | 2145 | |||
NM_001130153.1 | NP_001123625.1 | 21 | 2136 | |||
NM_008488.2 | NP_032514.1 | 21 | 2133 | |||
GenBank mRNA | AK012156.1 | - | 21 | 2087 | ||
AK043027.1 | - | 22 | 2269 | |||
AK051518.1 | - | 21 | 2087 | |||
AK131181.1 | - | 21 | 2033 | |||
AK157056.1 | - | 23 | 2249 | |||
AK172354.1 | - | 23 | 2249 | |||
BC012488.1 | - | 21 | 2096 | 3154558 | ||
U58203.1 | - | 21 | 2063 |
