siRNA Details
| Gene Symbol: Lcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010696.3 | NP_034826.2 | 20, 21 | 1643 | ||
| GenBank mRNA | AK036727.1 | - | 20, 21 | 1701 | ||
| AK153374.1 | - | 20, 21 | 1648 | |||
| AK168134.1 | - | 20, 21 | 1658 | |||
| AK170491.1 | - | 20, 21 | 1920 | |||
| BC006948.1 | - | 20, 21 | 1643 | 3603181 | ||
| U20159.1 | - | 20, 21 | 1682 | |||

Lcp2
Genomic Map